Merck
  • Use of Patient Health Records to Quantify Drug-Related Pro-arrhythmic Risk.
Use of Patient Health Records to Quantify Drug-Related Pro-arrhythmic Risk.
Cell reports. Medicine (2020-11-19)
Mark R Davies, Michael Martinec, Robert Walls, Roman Schwarz, Gary R Mirams, Ken Wang, Guido Steiner, Andy Surinach, Carlos Flores, Thierry Lavé, Thomas Singer, Liudmila Polonchuk
ABSTRACT
There is an increasing expectation that computational approaches may supplement existing human decision-making. Frontloading of models for cardiac safety prediction is no exception to this trend, and ongoing regulatory initiatives propose use of high-throughput in vitro data combined with computational models for calculating proarrhythmic risk. Evaluation of these models requires robust assessment of the outcomes. Using FDA Adverse Event Reporting System reports and electronic healthcare claims data from the Truven-MarketScan US claims database, we quantify the incidence rate of arrhythmia in patients and how this changes depending on patient characteristics. First, we propose that such datasets are a complementary resource for determining relative drug risk and assessing the performance of cardiac safety models for regulatory use. Second, the results suggest important determinants for appropriate stratification of patients and evaluation of additional drug risk in prescribing and clinical support algorithms and for precision health.
Product Description
USP
Risperidone, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Mexiletine hydrochloride, powder
Sigma-Aldrich
(±)-Verapamil hydrochloride, ≥99% (titration), powder
Sigma-Aldrich
Sodium hydroxide solution, BioUltra, for molecular biology, 10 M in H2O
Sigma-Aldrich
Nifedipine, Relatively selective blocker of L-type Ca2+ channels.
Sigma-Aldrich
Haloperidol, powder
Sigma-Aldrich
Clozapine
USP
Fluoxetine hydrochloride, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Pentamidine isethionate salt, powder
Sigma-Aldrich
Terfenadine
Sigma-Aldrich
(R)-(−)-Phenylephrine hydrochloride, powder
Sigma-Aldrich
(+)-cis-Diltiazem hydrochloride, ≥99% (HPLC)
Sigma-Aldrich
Nitrendipine, >95%, powder
Sigma-Aldrich
Pimozide
Sigma-Aldrich
Thioridazine hydrochloride, ≥99%
Sigma-Aldrich
Tamoxifen, ≥99%
Sigma-Aldrich
Imatinib mesylate, ≥98% (HPLC)
USP
Citalopram hydrobromide, United States Pharmacopeia (USP) Reference Standard