Merck
  • Cell Type-Specific Expression Analysis of the Inner Ear: A Technical Report.
Cell Type-Specific Expression Analysis of the Inner Ear: A Technical Report.
The Laryngoscope (2020-06-25)
Ronna Hertzano, Kathleen Gwilliam, Kevin Rose, Beatrice Milon, Maggie S Matern
ABSTRACT
The cellular diversity of the inner ear has presented a technical challenge in obtaining molecular insight into its development and function. The application of technological advancements in cell type-specific expression enable clinicians and researchers to leap forward from classic genetics to obtaining mechanistic understanding of congenital and acquired hearing loss. This understanding is essential for development of therapeutics to prevent and reverse diseases of the inner ear, including hearing loss. The objective of this study is to describe and compare the available tools for cell type-specific analysis of the ear, as a means to support decision making in study design. Three major approaches for cell type-specific analysis of the ear including fluorescence-activated cell sorting (FACS), ribosomal and RNA pulldown techniques, and single cell RNA-seq (scRNA-seq) are compared and contrasted using both published and original data. We demonstrate the strength and weaknesses of these approaches leading to the inevitable conclusion that to maximize the utility of these approaches, it is important to match the experimental approach with the tissue of origin, cell type of interest, and the biological question. Often, a combined approach (eg, cell sorting and scRNA-seq or expression analysis using 2 separate approaches) is required. Finally, new tools for visualization and analysis of complex expression data, such as the gEAR platform (umgear.org), collate cell type-specific gene expression from the ear field and provide unprecedented access to both clinicians and researchers. N/A Laryngoscope, 131:S1-S16, 2021.
Product Description
SAFC
Iscove′s Modified Dulbecco′s Medium, With sodium bicarbonate and L-glutamine, liquid, sterile-filtered, suitable for cell culture, suitable for hybridoma
Sigma-Aldrich
Protease from Bacillus licheniformis, Type VIII, lyophilized powder, 7-15 units/mg solid
Sigma-Aldrich
Accutase® solution, sterile-filtered, suitable for cell culture
Sigma-Aldrich
Thermolysin from Geobacillus stearothermophilus, powder, BioReagent, 30-350 units/mg protein (E1%/280), suitable for cell culture
Roche
Anti-Digoxigenin-AP, Fab fragments, from sheep
Roche
BM-Purple, Roche, pkg of 100 mL, solution