Merck
  • PDCD4 regulates axonal growth by translational repression of neurite growth-related genes and is modulated during nerve injury responses.
PDCD4 regulates axonal growth by translational repression of neurite growth-related genes and is modulated during nerve injury responses.
RNA (New York, N.Y.) (2020-08-05)
Andrés Di Paolo, Guillermo Eastman, Raquel Mesquita-Ribeiro, Joaquina Farias, Andrew Macklin, Thomas Kislinger, Nancy Colburn, David Munroe, José R Sotelo Sosa, Federico Dajas-Bailador, José R Sotelo-Silveira
ABSTRACT
Programmed cell death 4 (PDCD4) protein is a tumor suppressor that inhibits translation through the mTOR-dependent initiation factor EIF4A, but its functional role and mRNA targets in neurons remain largely unknown. Our work identified that PDCD4 is highly expressed in axons and dendrites of CNS and PNS neurons. Using loss- and gain-of-function experiments in cortical and dorsal root ganglia primary neurons, we demonstrated the capacity of PDCD4 to negatively control axonal growth. To explore PDCD4 transcriptome and translatome targets, we used Ribo-seq and uncovered a list of potential targets with known functions as axon/neurite outgrowth regulators. In addition, we observed that PDCD4 can be locally synthesized in adult axons in vivo, and its levels decrease at the site of peripheral nerve injury and before nerve regeneration. Overall, our findings demonstrate that PDCD4 can act as a new regulator of axonal growth via the selective control of translation, providing a target mechanism for axon regeneration and neuronal plasticity processes in neurons.
Sigma-Aldrich
Sodium citrate tribasic dihydrate, ACS reagent, ≥99.0%
Millipore
Benzonase® Nuclease, ≥250 units/μL, ≥90% (SDS-PAGE), recombinant, expressed in E. coli, buffered aqueous glycerol solution
Sigma-Aldrich
Antibiotic Antimycotic Solution (100×), Stabilized, with 10,000 units penicillin, 10 mg streptomycin and 25 μg amphotericin B per mL, 0.1 μm filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Paraformaldehyde, powder, 95%
Sigma-Aldrich
Puromycin dihydrochloride from Streptomyces alboniger, ≥98% (HPLC), powder
Sigma-Aldrich
Cycloheximide, ≥90% (HPLC)