Merck
  • Collagen hydrogels loaded with fibroblast growth factor-2 as a bridge to repair brain vessels in organotypic brain slices.
Collagen hydrogels loaded with fibroblast growth factor-2 as a bridge to repair brain vessels in organotypic brain slices.
Experimental brain research (2020-08-31)
Buket Ucar, Sedef Yusufogullari, Christian Humpel
ABSTRACT
Vessel damage is a general pathological process in many neurodegenerative disorders, as well as spinal cord injury, stroke, or trauma. Biomaterials can present novel tools to repair and regenerate damaged vessels. The aim of the present study is to test collagen hydrogels loaded with different angiogenic factors to study vessel repair in organotypic brain slice cultures. In the experimental set up I, we made a cut on the organotypic brain slice and tested re-growth of laminin + vessels. In the experimental set up II, we cultured two half brain slices with a gap with a collagen hydrogel placed in between to study endothelial cell migration. In the experimental set up I, we showed that the number of vessels crossing the cut was tendencially increased with the addition of fibroblast growth factor-2 (FGF-2), vascular endothelial growth factor, or platelet-derived growth factor-BB compared to the control group. In the experimental set up II, we demonstrated that a collagen hydrogel loaded with FGF-2 resulted in a significantly increased number of migrated laminin + cells in the gap between the slices compared to the control hydrogel. Co-administration of several growth factors did not further potentiate the effects. Taken together, we show that organotypic brain slices are good models to study brain vessels and FGF-2 is a potent angiogenic factor for endothelial cell proliferation and migration. Our results provide evidence that the collagen hydrogels can be used as an extracellular matrix for the vascular endothelial cells.
Product Description
Millipore
Polycarbonate Membrane Filter, 0.4 μm Pore Size, Isopore, filter diam. 25 mm, hydrophilic, white
Sigma-Aldrich
Anti-Laminin antibody produced in rabbit, 0.5 mg/mL, affinity isolated antibody, buffered aqueous solution