Merck
  • In vitro activities of natural products against oral Candida isolates from denture wearers.
In vitro activities of natural products against oral Candida isolates from denture wearers.
BMC complementary and alternative medicine (2011-11-29)
Cristina Marcos-Arias, Elena Eraso, Lucila Madariaga, Guillermo Quindós
ABSTRACT
Candida-associated denture stomatitis is a frequent infectious disease. Treatment of this oral condition is difficult because failures and recurrences are common. The aim of this study was to test the in vitro antifungal activity of pure constituents of essentials oils. Eight terpenic derivatives (carvacrol, farnesol, geraniol, linalool, menthol, menthone, terpinen-4-ol, and α-terpineol), a phenylpropanoid (eugenol), a phenethyl alcohol (tyrosol) and fluconazole were evaluated against 38 Candida isolated from denture-wearers and 10 collection Candida strains by the CLSI M27-A3 broth microdilution method. Almost all the tested compounds showed antifungal activity with MIC ranges of 0.03-0.25% for eugenol and linalool, 0.03-0.12% for geraniol, 0.06-0.5% for menthol, α-terpineol and terpinen-4-ol, 0.03-0.5% for carvacrol, and 0.06-4% for menthone. These compounds, with the exception of farnesol, menthone and tyrosol, showed important in vitro activities against the fluconazole-resistant and susceptible-dose dependent Candida isolates. Carvacrol, eugenol, geraniol, linalool and terpinen-4-ol were very active in vitro against oral Candida isolates. Their fungistatic and fungicidal activities might convert them into promising alternatives for the topic treatment of oral candidiasis and denture stomatitis.
Sigma-Aldrich
TWEEN® 80, BioXtra, viscous liquid
Sigma-Aldrich
TWEEN® 80, suitable for cell culture, suitable for insect cell culture, viscous liquid
Sigma-Aldrich
TWEEN® 80, viscous liquid, Low-peroxide
Sigma-Aldrich
TWEEN® 80, viscous liquid, Preservative Free, Low-peroxide; Low-carbonyls
Sigma-Aldrich
TWEEN® 80, from non-animal source
Sigma-Aldrich
TWEEN® 80, for molecular biology, syrup
Sigma-Aldrich
TWEEN® 80, viscous liquid
Sigma-Aldrich
ECO TWEEN® 80, viscous liquid