Merck
  • Phosphorylation of PDHA by AMPK Drives TCA Cycle to Promote Cancer Metastasis.
Phosphorylation of PDHA by AMPK Drives TCA Cycle to Promote Cancer Metastasis.
Molecular cell (2020-10-07)
Zhen Cai, Chien-Feng Li, Fei Han, Chunfang Liu, Anmei Zhang, Che-Chia Hsu, Danni Peng, Xian Zhang, Guoxiang Jin, Abdol-Hossein Rezaeian, Guihua Wang, Weina Zhang, Bo-Syong Pan, Chi-Yun Wang, Yu-Hui Wang, Shih-Ying Wu, Shun-Chin Yang, Fang-Chi Hsu, Ralph B D'Agostino, Christina M Furdui, Gregory L Kucera, John S Parks, Floyd H Chilton, Chih-Yang Huang, Fuu-Jen Tsai, Boris Pasche, Kounosuke Watabe, Hui-Kuan Lin
ABSTRACT
Cancer metastasis accounts for the major cause of cancer-related deaths. How disseminated cancer cells cope with hostile microenvironments in secondary site for full-blown metastasis is largely unknown. Here, we show that AMPK (AMP-activated protein kinase), activated in mouse metastasis models, drives pyruvate dehydrogenase complex (PDHc) activation to maintain TCA cycle (tricarboxylic acid cycle) and promotes cancer metastasis by adapting cancer cells to metabolic and oxidative stresses. This AMPK-PDHc axis is activated in advanced breast cancer and predicts poor metastasis-free survival. Mechanistically, AMPK localizes in the mitochondrial matrix and phosphorylates the catalytic alpha subunit of PDHc (PDHA) on two residues S295 and S314, which activates the enzymatic activity of PDHc and alleviates an inhibitory phosphorylation by PDHKs, respectively. Importantly, these phosphorylation events mediate PDHc function in cancer metastasis. Our study reveals that AMPK-mediated PDHA phosphorylation drives PDHc activation and TCA cycle to empower cancer cells adaptation to metastatic microenvironments for metastasis.
Sigma-Aldrich
Anti-PDK2 antibody produced in rabbit, affinity isolated antibody
Sigma-Aldrich
Monoclonal Anti-β-Actin antibody produced in mouse, clone AC-15, ascites fluid
Sigma-Aldrich
Monoclonal ANTI-FLAG® M2 antibody produced in mouse, 1 mg/mL, clone M2, affinity isolated antibody, buffered aqueous solution (50% glycerol, 10 mM sodium phosphate, and 150 mM NaCl, pH 7.4)