Merck
  • Phosphoproteomic Analysis across the Yeast Life Cycle Reveals Control of Fatty Acyl Chain Length by Phosphorylation of the Fatty Acid Synthase Complex.
Phosphoproteomic Analysis across the Yeast Life Cycle Reveals Control of Fatty Acyl Chain Length by Phosphorylation of the Fatty Acid Synthase Complex.
Cell reports (2020-08-14)
Fernando Martínez-Montañés, Albert Casanovas, Richard R Sprenger, Magdalena Topolska, David L Marshall, Marta Moreno-Torres, Berwyck L J Poad, Stephen J Blanksby, Martin Hermansson, Ole N Jensen, Christer S Ejsing
ABSTRACT
The ability to remodel lipid metabolism under changing conditions is pivotal for cellular functionality and homeostasis. Here, we characterize the regulatory landscape of phosphorylation-based signaling events across the life cycle of Saccharomyces cerevisiae and determine its impact on the regulation of lipid metabolism. Our data show that 50 lipid metabolic proteins are differentially phosphorylated as cells transit between different physiological states. To identify functional phosphosites, we devised a strategy where multiple phosphosites are simultaneously mutated into phosphomimetic or phosphodeficient alleles and mutants are phenotyped by in-depth lipidomics flux analysis. This uncovers functional phosphosites in the phosphatidate cytidylyltransferase Cds1, the phosphatidylserine synthase Cho1, and Fas2, the α-subunit of the fatty acid synthase (FAS) complex. Furthermore, we show that the fatty acyl chain length produced by FAS is governed by phosphorylation. Overall, our work demonstrates a vital role for phosphoregulation of lipid metabolism and provides a resource to investigate its molecular underpinnings.
Sigma-Aldrich
Hydrochloric acid, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., fuming, ≥37%, APHA: ≤10
Sigma-Aldrich
Lithium acetate, 99.95% trace metals basis
Sigma-Aldrich
MS PhosphoMix 1 Light, Phosphopeptide Standard for MS
Sigma-Aldrich
MS PhosphoMix 1 Heavy, Phosphopeptide Standard for MS
Sigma-Aldrich
Malonyl-13C3 coenzyme A lithium salt, 99 atom % 13C, 95% (CP)
Supelco
Tris(2-carboxyethyl)phosphine hydrochloride solution, 0.5 M, pH 7.0(aqueous solution; pH was adjusted with ammonium hydroxide)
Sigma-Aldrich
MS PhosphoMix 2 Heavy, Phosphopeptide Standard for MS
Sigma-Aldrich
MS PhosphoMix 2 Light, Phosphopeptide Standard for MS
Sigma-Aldrich
L-Cysteine, 97%
Sigma-Aldrich
Sodium deoxycholate monohydrate, BioUltra, ≥99.0% (NT)
Sigma-Aldrich
MS PhosphoMix 3 Light, Phosphopeptide Standard for MS