Merck
  • EDEM1 Drives Misfolded Protein Degradation via ERAD and Exploits ER-Phagy as Back-Up Mechanism When ERAD Is Impaired.
EDEM1 Drives Misfolded Protein Degradation via ERAD and Exploits ER-Phagy as Back-Up Mechanism When ERAD Is Impaired.
International journal of molecular sciences (2020-05-20)
Marioara Chiritoiu, Gabriela N Chiritoiu, Cristian V A Munteanu, Florin Pastrama, N Erwin Ivessa, Stefana M Petrescu
ABSTRACT
Endoplasmic reticulum (ER)-associated degradation (ERAD) is the main mechanism of targeting ER proteins for degradation to maintain homeostasis, and perturbations of ERAD lead to pathological conditions. ER-degradation enhancing α-mannosidase-like (EDEM1) was proposed to extract terminally misfolded proteins from the calnexin folding cycle and target them for degradation by ERAD. Here, using mass-spectrometry and biochemical methods, we show that EDEM1 is found in auto-regulatory complexes with ERAD components. Moreover, the N-terminal disordered region of EDEM1 mediates protein-protein interaction with misfolded proteins, whilst the absence of this domain significantly impairs their degradation. We also determined that overexpression of EDEM1 can induce degradation, even when proteasomal activity is severely impaired, by promoting the formation of aggregates, which can be further degraded by autophagy. Therefore, we propose that EDEM1 maintains ER homeostasis and mediates ERAD client degradation via autophagy when either dislocation or proteasomal degradation are impaired.
Sigma-Aldrich
MISSION® esiRNA, targeting human OS9
Sigma-Aldrich
Ethenesulfonyl fluoride, 95%
Sigma-Aldrich
Z-Leu-Leu-Leu-al, ≥90% (HPLC)
Sigma-Aldrich
MISSION® esiRNA, targeting human SEL1L
Sigma-Aldrich
MISSION® esiRNA, targeting human SYVN1
Sigma-Aldrich
MISSION® esiRNA, targeting human VCP
Sigma-Aldrich
DL-Cysteine, technical grade