Merck
  • Neuromodulator Signaling Bidirectionally Controls Vesicle Numbers in Human Synapses.
Neuromodulator Signaling Bidirectionally Controls Vesicle Numbers in Human Synapses.
Cell (2019-10-05)
Christopher Patzke, Marisa M Brockmann, Jinye Dai, Kathlyn J Gan, M Katharina Grauel, Pascal Fenske, Yu Liu, Claudio Acuna, Christian Rosenmund, Thomas C Südhof
ABSTRACT
Neuromodulators bind to pre- and postsynaptic G protein-coupled receptors (GPCRs), are able to quickly change intracellular cyclic AMP (cAMP) and Ca2+ levels, and are thought to play important roles in neuropsychiatric and neurodegenerative diseases. Here, we discovered in human neurons an unanticipated presynaptic mechanism that acutely changes synaptic ultrastructure and regulates synaptic communication. Activation of neuromodulator receptors bidirectionally controlled synaptic vesicle numbers within nerve terminals. This control correlated with changes in the levels of cAMP-dependent protein kinase A-mediated phosphorylation of synapsin-1. Using a conditional deletion approach, we reveal that the neuromodulator-induced control of synaptic vesicle numbers was largely dependent on synapsin-1. We propose a mechanism whereby non-phosphorylated synapsin-1 "latches" synaptic vesicles to presynaptic clusters at the active zone. cAMP-dependent phosphorylation of synapsin-1 then removes the vesicles. cAMP-independent dephosphorylation of synapsin-1 in turn recruits vesicles. Synapsin-1 thereby bidirectionally regulates synaptic vesicle numbers and modifies presynaptic neurotransmitter release as an effector of neuromodulator signaling in human neurons.
Sigma-Aldrich
Anti-BSN antibody produced in rabbit, 1 mg/mL, affinity isolated antibody
Sigma-Aldrich
Anti-NeuN Antibody (rabbit), from rabbit, purified by affinity chromatography
Sigma-Aldrich
Anti-Adrenergic Receptor α-2A antibody produced in rabbit, affinity isolated antibody
Sigma-Aldrich
Anti-β-Tubulin III antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
Sigma-Aldrich
Puromycin dihydrochloride from Streptomyces alboniger, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Cytosine β-D-arabinofuranoside hydrochloride, crystalline