Merck
  • Plasticity of nuclear and cytoplasmic stress responses of RNA-binding proteins.
Plasticity of nuclear and cytoplasmic stress responses of RNA-binding proteins.
Nucleic acids research (2020-04-22)
Michael Backlund, Frank Stein, Mandy Rettel, Thomas Schwarzl, Joel I Perez-Perri, Annika Brosig, Yang Zhou, Gabriele Neu-Yilik, Matthias W Hentze, Andreas E Kulozik
ABSTRACT
Cellular stress causes multifaceted reactions to trigger adaptive responses to environmental cues at all levels of the gene expression pathway. RNA-binding proteins (RBP) are key contributors to stress-induced regulation of RNA fate and function. Here, we uncover the plasticity of the RNA interactome in stressed cells, differentiating between responses in the nucleus and in the cytoplasm. We applied enhanced RNA interactome capture (eRIC) analysis preceded by nucleo-cytoplasmic fractionation following arsenite-induced oxidative stress. The data reveal unexpectedly compartmentalized RNA interactomes and their responses to stress, including differential responses of RBPs in the nucleus versus the cytoplasm, which would have been missed by whole cell analyses.
Sigma-Aldrich
Ribonuclease T1 from Aspergillus oryzae, ammonium sulfate suspension, 300,000-600,000 units/mg protein
Sigma-Aldrich
MES, low moisture content, ≥99% (titration)
Sigma-Aldrich
Monoclonal Anti-α-Tubulin antibody produced in mouse, ascites fluid, clone B-5-1-2
Sigma-Aldrich
Lithium dodecyl sulfate, BioReagent, for molecular biology, suitable for electrophoresis
Sigma-Aldrich
N-(3-Dimethylaminopropyl)-N′-ethylcarbodiimide hydrochloride, commercial grade, powder
Roche
cOmplete, EDTA-free Protease Inhibitor Cocktail, Tablets provided in EASYpacks
Sigma-Aldrich
Ethanolamine, ≥98%
Sigma-Aldrich
Anti-Mouse IgG (whole molecule)–Peroxidase antibody produced in rabbit, IgG fraction of antiserum, buffered aqueous solution
Sigma-Aldrich
Anti-Rabbit IgG (whole molecule)–Peroxidase antibody produced in goat, affinity isolated antibody
Sigma-Aldrich
Ribonuclease A from bovine pancreas, (Solution of 50% glycerol, 10mM Tris-HCL pH 8.0)