Merck
Use of Protein Folding Reagents.
Current protocols in protein science (2016-04-03)
ABSTRACT
The reagents and methods for purification and use of the most commonly used denaturants, guanidine hydrochloride (guanidine-HCl) and urea, are described. Other protein denaturants and reagents used to fold proteins are briefly mentioned. Sulfhydryl reagents (reducing agents) and "oxido-shuffling" (or oxidative regeneration) systems are also described.
Sigma-Aldrich
Guanidine hydrochloride, for molecular biology, ≥99%
Sigma-Aldrich
Guanidine hydrochloride, BioUltra, for molecular biology, ≥99.5% (AT)
Sigma-Aldrich
Guanidine hydrochloride, organic base and chaeotropic agent, ≥99% (titration)