Merck
  • Fluorescent Antibiotics: New Research Tools to Fight Antibiotic Resistance.
Fluorescent Antibiotics: New Research Tools to Fight Antibiotic Resistance.
Trends in biotechnology (2018-02-27)
M Rhia L Stone, Mark S Butler, Wanida Phetsang, Matthew A Cooper, Mark A T Blaskovich
ABSTRACT
Better understanding how multidrug-resistant (MDR) bacteria can evade current and novel antibiotics requires a better understanding of the chemical biology of antibiotic action. This necessitates using new tools and techniques to advance our knowledge of bacterial responses to antibiotics, ideally in live cells in real time, to selectively investigate bacterial growth, division, metabolism, and resistance in response to antibiotic challenge. In this review, we discuss the preparation and biological evaluation of fluorescent antibiotics, focussing on how these reporters and assay methods can help elucidate resistance mechanisms. We also examine the potential utility of such probes for real-time in vivo diagnosis of infections.
Sigma-Aldrich
Rhodamine B labeled Ramoplanin, For fluorescent microbial imaging
Sigma-Aldrich
FITC labeled vancomycin, for fluorescent microbial imaging
Sigma-Aldrich
Dansyl labeled polymyxin B Ready Made Solution, for fluorescent microbial imaging, 1.5 mg/mL in H2O
Sigma-Aldrich
Rhodamine B labeled Polymyxin B Ready Made Solution, For fluorescent microbial imaging, 1.1 mg/mL in water