Merck
  • PD-1 and BTLA regulate T cell signaling differentially and only partially through SHP1 and SHP2.
PD-1 and BTLA regulate T cell signaling differentially and only partially through SHP1 and SHP2.
The Journal of cell biology (2020-05-22)
Xiaozheng Xu, Bowen Hou, Amitkumar Fulzele, Takeya Masubuchi, Yunlong Zhao, Zijun Wu, Yanyan Hu, Yong Jiang, Yanzhe Ma, Haopeng Wang, Eric J Bennett, Guo Fu, Enfu Hui
ABSTRACT
Blockade antibodies of the immunoinhibitory receptor PD-1 can stimulate the anti-tumor activity of T cells, but clinical benefit is limited to a fraction of patients. Evidence suggests that BTLA, a receptor structurally related to PD-1, may contribute to resistance to PD-1 targeted therapy, but how BTLA and PD-1 differ in their mechanisms is debated. Here, we compared the abilities of BTLA and PD-1 to recruit effector molecules and to regulate T cell signaling. While PD-1 selectively recruited SHP2 over the stronger phosphatase SHP1, BTLA preferentially recruited SHP1 to more efficiently suppress T cell signaling. Contrary to the dominant view that PD-1 and BTLA signal exclusively through SHP1/2, we found that in SHP1/2 double-deficient primary T cells, PD-1 and BTLA still potently inhibited cell proliferation and cytokine production, albeit more transiently than in wild type T cells. Thus, PD-1 and BTLA can suppress T cell signaling through a mechanism independent of both SHP1 and SHP2.
Sigma-Aldrich
Imidazole, ReagentPlus®, 99%
Sigma-Aldrich
PMA, for use in molecular biology applications, ≥99% (HPLC)
Avanti
18:1 PEG5000 PE, Avanti Research - A Croda Brand 880230C
Avanti
18:1 PEG5000 PE, Avanti Research - A Croda Brand 880230P, powder
Avanti
16:0-18:1 PC, Avanti Research - A Croda Brand
Avanti
16:0-18:1 PC, Avanti Research - A Croda Brand
Sigma-Aldrich
1,2-Dioleoyl-sn-glycero-3-phosphocholine, lyophilized powder
Sigma-Aldrich
L-Glutathione reduced, ≥98.0%
BRAND® 96-well microplate, U-bottom, round bottom, non-sterile
Avanti
16:0 Liss Rhod PE, Avanti Research - A Croda Brand 810158C
Avanti
16:0 Liss Rhod PE, Avanti Research - A Croda Brand
Avanti
18:1 DGS-NTA(Ni), Avanti Research - A Croda Brand
Avanti
18:1 DGS-NTA(Ni), Avanti Research - A Croda Brand
Sigma-Aldrich
Ionomycin, Free Acid, Streptomyces conglobatus, Ionomycin, Free Acid, Streptomyces conglobatus, CAS 56092-81-0, is a highly specific for divalent cations and more effective than A23187 (Cat. No. 100105) as a mobile ion carrier for Ca2+.