Merck
  • Propionic Acid Shapes the Multiple Sclerosis Disease Course by an Immunomodulatory Mechanism.
Propionic Acid Shapes the Multiple Sclerosis Disease Course by an Immunomodulatory Mechanism.
Cell (2020-03-12)
Alexander Duscha, Barbara Gisevius, Sarah Hirschberg, Nissan Yissachar, Gabriele I Stangl, Eva Eilers, Verian Bader, Stefanie Haase, Johannes Kaisler, Christina David, Ruth Schneider, Riccardo Troisi, Daniel Zent, Tobias Hegelmaier, Nikolaos Dokalis, Sara Gerstein, Sara Del Mare-Roumani, Sivan Amidror, Ori Staszewski, Gereon Poschmann, Kai Stühler, Frank Hirche, Andras Balogh, Stefan Kempa, Pascal Träger, Mario M Zaiss, Jacob Bak Holm, Megan G Massa, Henrik Bjørn Nielsen, Andreas Faissner, Carsten Lukas, Sören G Gatermann, Markus Scholz, Horst Przuntek, Marco Prinz, Sofia K Forslund, Konstanze F Winklhofer, Dominik N Müller, Ralf A Linker, Ralf Gold, Aiden Haghikia
ABSTRACT
Short-chain fatty acids are processed from indigestible dietary fibers by gut bacteria and have immunomodulatory properties. Here, we investigate propionic acid (PA) in multiple sclerosis (MS), an autoimmune and neurodegenerative disease. Serum and feces of subjects with MS exhibited significantly reduced PA amounts compared with controls, particularly after the first relapse. In a proof-of-concept study, we supplemented PA to therapy-naive MS patients and as an add-on to MS immunotherapy. After 2 weeks of PA intake, we observed a significant and sustained increase of functionally competent regulatory T (Treg) cells, whereas Th1 and Th17 cells decreased significantly. Post-hoc analyses revealed a reduced annual relapse rate, disability stabilization, and reduced brain atrophy after 3 years of PA intake. Functional microbiome analysis revealed increased expression of Treg-cell-inducing genes in the intestine after PA intake. Furthermore, PA normalized Treg cell mitochondrial function and morphology in MS. Our findings suggest that PA can serve as a potent immunomodulatory supplement to MS drugs.
Sigma-Aldrich
2-Ethylbutyric acid, ≥98%, FCC, FG
Sigma-Aldrich
Dulbecco′s Modified Eagle′s Medium, Without glucose, L-glutamine, phenol red, sodium pyruvate and sodium bicarbonate, powder, suitable for cell culture
Sigma-Aldrich
4-Guanidinobutyric acid, ≥98%
Sigma-Aldrich
Sodium pyruvate, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥99%
Sigma-Aldrich
2-Ethylbutyric acid, 99%
Sigma-Aldrich
Phorbol 12-myristate 13-acetate, ≥99% (TLC), film or powder
Sigma-Aldrich
Glycine ethyl ester hydrochloride, 99%
Sigma-Aldrich
Sodium butyrate-4-13C, 99 atom % 13C
Sigma-Aldrich
Rotenone, ≥95%
Sigma-Aldrich
Antimycin A from Streptomyces sp.
Sigma-Aldrich
Sodium butyrate, 98%
Sigma-Aldrich
Crotonic acid, 98%
Sigma-Aldrich
Perchloric acid, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., 70.0-72.0%