Merck
  • Different regions of synaptic vesicle membrane regulate VAMP2 conformation for the SNARE assembly.
Different regions of synaptic vesicle membrane regulate VAMP2 conformation for the SNARE assembly.
Nature communications (2020-03-27)
Chuchu Wang, Jia Tu, Shengnan Zhang, Bin Cai, Zhenying Liu, Shouqiao Hou, Qinglu Zhong, Xiao Hu, Wenbin Liu, Guohui Li, Zhijun Liu, Lin He, Jiajie Diao, Zheng-Jiang Zhu, Dan Li, Cong Liu
ABSTRACT
Vesicle associated membrane protein 2 (VAMP2/synaptobrevin2), a core SNARE protein residing on synaptic vesicles (SVs), forms helix bundles with syntaxin-1 and SNAP25 for the SNARE assembly. Prior to the SNARE assembly, the structure of VAMP2 is unclear. Here, by using in-cell NMR spectroscopy, we describe the dynamic membrane association of VAMP2 SNARE motif in mammalian cells, and the structural change of VAMP2 upon the change of intracellular lipid environment. We analyze the lipid compositions of the SV membrane by mass-spectrometry-based lipidomic profiling, and further reveal that VAMP2 forms distinctive conformations in different membrane regions. In contrast to the non-raft region, the membrane region of cholesterol-rich lipid raft markedly weakens the membrane association of VAMP2 SNARE motif, which releases the SNARE motif and facilitates the SNARE assembly. Our work reveals the regulation of different membrane regions on VAMP2 structure and sheds light on the spatial regulation of SNARE assembly.
Sigma-Aldrich
Monoclonal Anti-Synaptophysin antibody produced in mouse, clone SVP-38, ascites fluid
Sigma-Aldrich
Mevastatin, ≥98% (HPLC), powder or crystals
Avanti
15:0-18:1-d7-PI, Avanti Research - A Croda Brand
Avanti
15:0-18:1-d7-PE, Avanti Research - A Croda Brand
Avanti
18:1-d7 Lyso PC, Avanti Research - A Croda Brand
Avanti
15:0-18:1-d7-PS, Avanti Research - A Croda Brand
Avanti
18:1-d7 Lyso PE, Avanti Research - A Croda Brand 791644C
Avanti
15:0-18:1-d7-PG, Avanti Research - A Croda Brand 791640C
Avanti
15:0-18:1-d7-PC, Avanti Research - A Croda Brand
Avanti
18:1-d9 SM, Avanti Research - A Croda Brand