Merck
  • N-acylhomoserine lactone regulates violacein production in Chromobacterium violaceum type strain ATCC 12472.
N-acylhomoserine lactone regulates violacein production in Chromobacterium violaceum type strain ATCC 12472.
FEMS microbiology letters (2008-01-08)
Tomohiro Morohoshi, Masashi Kato, Katsumasa Fukamachi, Norihiro Kato, Tsukasa Ikeda
ABSTRACT
In tests, Chromobacterium violaceum ATCC 12472 produced several N-acyl-L-homoserine lactones (AHLs). Of these, N-(3-hydroxydecanoyl)-L-homoserine lactone was dominant, and controlled violacein production by quorum sensing. Strain VIR07, an AHL-deficient mutant, did not produce violacein. Violacein production in VIR07 was induced by adding long-chain AHLs (C10-C16), but was inhibited by adding short-chain AHLs (C4-C8). Strain VIR07 showed the response of violacein production when AHLs diffused from a variety of AHL-producing bacteria on agar plates, and thus might be a useful biosensor for recognizing exogenous AHLs.
Sigma-Aldrich
N-Dodecanoyl-DL-homoserine lactone, ≥97.0% (HPLC)
Sigma-Aldrich
N-Octanoyl-DL-homoserine lactone, ≥97.0% (HPLC)
Sigma-Aldrich
N-Tetradecanoyl-DL-homoserine lactone, ≥97.0% (HPLC)
Sigma-Aldrich
N-Butyryl-DL-homoserine lactone, ≥96.0% (HPLC)
Sigma-Aldrich
N-Hexanoyl-DL-homoserine lactone, ≥97.0% (HPLC)
Sigma-Aldrich
N-Decanoyl-DL-homoserine lactone, ≥97.0% (HPLC)
Sigma-Aldrich
N-Heptanoyl-DL-homoserine lactone, ≥97.0% (HPLC)