Merck
  • Identification and Characterization of ShSPI, a Kazal-Type Elastase Inhibitor from the Venom of Scolopendra Hainanum.
Identification and Characterization of ShSPI, a Kazal-Type Elastase Inhibitor from the Venom of Scolopendra Hainanum.
Toxins (2019-12-11)
Ning Luan, Qiyu Zhao, Zilei Duan, Mengyao Ji, Meichen Xing, Tengyu Zhu, James Mwangi, Mingqiang Rong, Jiangxin Liu, Ren Lai
ABSTRACT
Elastase is a globular glycoprotein and belongs to the chymotrypsin family. It is involved in several inflammatory cascades on the basis of cleaving the important connective tissue protein elastin, and is strictly regulated to a balance by several endogenous inhibitors. When elastase and its inhibitors are out of balance, severe diseases will develop, especially those involved in the cardiopulmonary system. Much attention has been attracted in seeking innovative elastase inhibitors and various advancements have been taken on clinical trials of these inhibitors. Natural functional peptides from venomous animals have been shown to have anti-protease properties. Here, we identified a kazal-type serine protease inhibitor named ShSPI from the cDNA library of the venom glands of Scolopendra hainanum. ShSPI showed significant inhibitory effects on porcine pancreatic elastase and human neutrophils elastase with Ki values of 225.83 ± 20 nM and 12.61 ± 2 nM, respectively. Together, our results suggest that ShSPI may be an excellent candidate to develop a drug for cardiopulmonary diseases.
Sigma-Aldrich
Tris(2-carboxyethyl)phosphine hydrochloride, BioUltra, ≥98% (NMR)
Sigma-Aldrich
Nα-Benzoyl-L-arginine 4-nitroanilide hydrochloride, ≥99% (TLC), suitable for substrate for trypsin
Sigma-Aldrich
N-Succinyl-Gly-Gly-Phe-p-nitroanilide, protease substrate
Sigma-Aldrich
Thrombin generation chromogenic substrate, ≥90% (HPLC), solid
Millipore
Elastase Substrate I, Colorimetric, A specific chromogenic substrate for human neutrophil elastase.
Sigma-Aldrich
Factor Xa chromogenic substrate, solid
Sigma-Aldrich
Iodoacetamide, BioUltra
Sigma-Aldrich
Guanidine hydrochloride, organic base and chaeotropic agent, ≥99% (titration)
Sigma-Aldrich
Z-Phe-Arg 7-amido-4-methylcoumarin hydrochloride, kallikrein substrate
Sigma-Aldrich
Thrombin from human plasma, lyophilized powder, 1500-3500 NIH units/mg protein (E1%/280, 18.3), suitable for cell culture
Sigma-Aldrich
Kallikrein, Human Plasma, Native Kallikrein from human plasma. A serine protease that releases bradykinin from kininogen. Present in the plasma as an inactive precursor, prokallikrein.