Merck
  • Human Mincle Binds to Cholesterol Crystals and Triggers Innate Immune Responses.
Human Mincle Binds to Cholesterol Crystals and Triggers Innate Immune Responses.
The Journal of biological chemistry (2015-08-25)
Ryoko Kiyotake, Masatsugu Oh-Hora, Eri Ishikawa, Tomofumi Miyamoto, Tatsuro Ishibashi, Sho Yamasaki
ABSTRACT
C-type lectin receptors (CLRs) are an emerging family of pattern recognition receptors that recognizes pathogens or damaged tissue to trigger innate immune responses. However, endogenous ligands for CLRs are not fully understood. In this study, we sought to identify an endogenous ligand(s) for human macrophage-inducible C-type lectin (hMincle). A particular fraction of lipid extracts from liver selectively activated reporter cells expressing hMincle. MS analysis determined the chemical structure of the active component as cholesterol. Purified cholesterol in plate-coated and crystalized forms activates reporter cells expressing hMincle but not murine Mincle (mMincle). Cholesterol crystals are known to activate immune cells and induce inflammatory responses through lysosomal damage. However, direct innate immune receptors for cholesterol crystals have not been identified. Murine macrophages transfected with hMincle responded to cholesterol crystals by producing pro-inflammatory cytokines. Human dendritic cells expressed a set of inflammatory genes in response to cholesterol crystals, and this was inhibited by anti-human Mincle. Importantly, other related CLRs did not bind cholesterol crystals, whereas other steroids were not recognized by hMincle. These results suggest that cholesterol crystals are an endogenous ligand for hMincle and that they activate innate immune responses.
Product Description
Avanti
desmosterol, Avanti Research - A Croda Brand
Sigma-Aldrich
Testosterone, ≥98%
Sigma-Aldrich
Cortisone, ≥95%
Avanti
trihydroxycholestanoic acid, Avanti Research - A Croda Brand
Avanti
Liver Extract Total, Avanti Research - A Croda Brand
Avanti
Liver Extract Total, Avanti Research - A Croda Brand 181104P, powder
Sigma-Aldrich
β-Estradiol, ≥98%