Merck
  • Qualitative and quantitative characterization of protein-phosphoinositide interactions with liposome-based methods.
Qualitative and quantitative characterization of protein-phosphoinositide interactions with liposome-based methods.
Autophagy (2013-03-01)
Ricarda A Busse, Andreea Scacioc, Javier M Hernandez, Roswitha Krick, Milena Stephan, Andreas Janshoff, Michael Thumm, Karin Kühnel
ABSTRACT
We characterized phosphoinositide binding of the S. cerevisiae PROPPIN Hsv2 qualitatively with density flotation assays and quantitatively through isothermal titration calorimetry (ITC) measurements using liposomes. We discuss the design of these experiments and show with liposome flotation assays that Hsv2 binds with high specificity to both PtdIns3P and PtdIns(3,5)P 2. We propose liposome flotation assays as a more accurate alternative to the commonly used PIP strips for the characterization of phosphoinositide-binding specificities of proteins. We further quantitatively characterized PtdIns3P binding of Hsv2 with ITC measurements and determined a dissociation constant of 0.67 µM and a stoichiometry of 2:1 for PtdIns3P binding to Hsv2. PtdIns3P is crucial for the biogenesis of autophagosomes and their precursors. Besides the PROPPINs there are other PtdIns3P binding proteins with a link to autophagy, which includes the FYVE-domain containing proteins ZFYVE1/DFCP1 and WDFY3/ALFY and the PX-domain containing proteins Atg20 and Snx4/Atg24. The methods described could be useful tools for the characterization of these and other phosphoinositide-binding proteins.
Product Description
Avanti
18:1 PI(4,5)P2, Avanti Research - A Croda Brand
Avanti
Brain PE, Avanti Research - A Croda Brand 840022C
Avanti
Egg PC, Avanti Research - A Croda Brand
Avanti
Brain PE, Avanti Research - A Croda Brand 840022P, powder
Avanti
Egg PC, Avanti Research - A Croda Brand
Avanti
18:1 PI(3)P, Avanti Research - A Croda Brand
Sigma-Aldrich
Sodium cholate hydrate, from bovine and/or ovine bile, ≥99%
Avanti
18:1 PI(3,4,5)P3, 1,2-dioleoyl-sn-glycero-3-phospho-(1′-myo-inositol-3′,4′,5′-trisphosphate) (ammonium salt), powder
Avanti
Brain PS, Avanti Research - A Croda Brand
Avanti
Brain PI(4)P, Avanti Research - A Croda Brand 840045P, powder
Avanti
18:1 PI(5)P, 1,2-dioleoyl-sn-glycero-3-phospho-(1′-myo-inositol-5′-phosphate) (ammonium salt), powder
Avanti
Brain PI(4)P, Avanti Research - A Croda Brand 840045X
Avanti
Brain PS, Avanti Research - A Croda Brand
Avanti
18:1 PI(3,5)P2, 1,2-dioleoyl-sn-glycero-3-phospho-(1′-myo-inositol-3′,5′-bisphosphate) (ammonium salt), powder