Merck
  • Identification of regulatory Hck and PAI-2 proteins in the monocyte response to PEG-containing matrices.
Identification of regulatory Hck and PAI-2 proteins in the monocyte response to PEG-containing matrices.
Biomaterials (2009-05-16)
Sean T Zuckerman, James F Brown, Weiyuan J Kao
ABSTRACT
Mass spectrometry is a powerful proteomic tool enabling researchers to survey the global proteome of a cell. This technique has only recently been employed to investigate cell-material interactions. We had previously identified material scarcity and limited adherent cells as challenges facing mass spectrometric analysis of cell-material interactions. U937 adherent to tissue culture poly(styrene) was used as a model system for identifying proteins expressed by adherent monocytes and analyzed by HPLC coupled offline to MALDI-ToF/ToF (LC-MALDI). We identified 645 proteins from two cation fractions of crude U937 monocyte cell lysate. Forty three proteins of interest from the 645 were chosen based on literature searches for relevance to monocyte-material inflammation and wound healing. Proteins such as 40S ribosomal protein S19 and tyrosyl tRNA synthetase highlight the ability of LC-MALDI to identify proteins relevant to monocyte-material interactions that are currently unexplored. We used PEG-based semi-interpenetrating polymer networks and PEG-only hydrogels to investigate surface dependent effects on the Src family kinase Hck and plasminogen activator inhibitor-2 (PAI-2) using the pyrazolo pyrimidine small molecule inhibitor PP2 and exogenous urokinase plasminogen activator addition, respectively. Hck is well researched in cell adhesion while PAI-2 is virtually unknown in cell-material interactions. U937 on TCPS and PEG-only hydrogels secreted similar levels of inflammatory cytokines and gelatinase MMP-9. MCP-1 secretion from monocytes on PEG-only hydrogels was Hck independent in contrast to Hck-dependent MCP-1 secretion in U937 on TCPS. Overall, U937 adherent to sIPNs secrete low levels of soluble gelatinase MMP-9, IL-1beta, TNF-alpha, IL-6, and MCP-1 independent of Hck and PAI-2. This work demonstrates significant changes in surface dependent expression of proteins from monocytes adherent to PEG-based materials compared to TCPS.
Sigma-Aldrich
(±)-3-Benzyloxy-1,2-propanediol, ≥97.0% (HPLC)
Sigma-Aldrich
PP2, PP2, CAS 172889-27-9, is a potent, reversible, ATP-competitive, inhibitor of the Src family of protein tyrosine kinases (IC₅₀ = 4, 5, 5, &100 nM for p56lck, p59fynT, Hck, & Src, respectively).
Sigma-Aldrich
Urokinase-type Plasminogen Activator