Merck
  • A comprehensive model for assessment of liver stage therapies targeting Plasmodium vivax and Plasmodium falciparum.
A comprehensive model for assessment of liver stage therapies targeting Plasmodium vivax and Plasmodium falciparum.
Nature communications (2018-05-11)
Alison Roth, Steven P Maher, Amy J Conway, Ratawan Ubalee, Victor Chaumeau, Chiara Andolina, Stephen A Kaba, Amélie Vantaux, Malina A Bakowski, Richard Thomson Luque, Swamy Rakesh Adapa, Naresh Singh, Samantha J Barnes, Caitlin A Cooper, Mélanie Rouillier, Case W McNamara, Sebastian A Mikolajczak, Noah Sather, Benoît Witkowski, Brice Campo, Stefan H I Kappe, David E Lanar, François Nosten, Silas Davidson, Rays H Y Jiang, Dennis E Kyle, John H Adams
ABSTRACT
Malaria liver stages represent an ideal therapeutic target with a bottleneck in parasite load and reduced clinical symptoms; however, current in vitro pre-erythrocytic (PE) models for Plasmodium vivax and P. falciparum lack the efficiency necessary for rapid identification and effective evaluation of new vaccines and drugs, especially targeting late liver-stage development and hypnozoites. Herein we report the development of a 384-well plate culture system using commercially available materials, including cryopreserved primary human hepatocytes. Hepatocyte physiology is maintained for at least 30 days and supports development of P. vivax hypnozoites and complete maturation of P. vivax and P. falciparum schizonts. Our multimodal analysis in antimalarial therapeutic research identifies important PE inhibition mechanisms: immune antibodies against sporozoite surface proteins functionally inhibit liver stage development and ion homeostasis is essential for schizont and hypnozoite viability. This model can be implemented in laboratories in disease-endemic areas to accelerate vaccine and drug discovery research.
Sigma-Aldrich
Schneider′s Insect Medium, With L-glutamine, without calcium chloride and sodium bicarbonate, powder, suitable for insect cell culture
Sigma-Aldrich
McCoy′s 5A Medium, Modified, with L-glutamine and sodium bicarbonate, liquid, sterile-filtered, suitable for cell culture
Sigma-Aldrich
Tetramethylrhodamine methyl ester perchlorate, ≥95%