Skip to Content
AM
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Viresolve
®
Pro Shield H
Products
Product Category
instruments (2)
Brand
Millipore (6)
Product Line
Length (cm)
Length (in)
Surface Area cm²
Quality Segment
Color
Housing Material
Membrane Material
Filter Products
Showing 1-6 of 6
Compare
Product Number
Description
Pricing
VPPH103NB1
Viresolve
®
Pro Shield H,
Virus prefilter, Viresolve
®
Pro Shield H, Modus 1.3 format, filtration area 0.22 m²
Expand
View Pricing
VPMHKITNB9
Viresolve
®
Pro Shield H,
Virus prefilter, Viresolve
®
Pro Shield H, Micro kit, filtration area 3.1 cm²
Expand
View Pricing
VPPH101NB1
Viresolve
®
Pro Shield H,
Virus prefilter, Viresolve
®
Pro Shield H, Modus 1.1 format, filtration area 0.017 m²
Expand
View Pricing
VPPH102NB1
Viresolve
®
Pro Shield H,
Virus prefilter, Viresolve
®
Pro Shield H, Modus 1.2 format, filtration area 0.07 m²
Expand
View Pricing
VPPH202NB1
Viresolve
®
Pro Shield H,
Virus prefilter, Viresolve
®
Pro Shield H, Magnus 2.2 format, filtration area 1.53 m²
Expand
View Pricing
VPPH201NB1
Viresolve
®
Pro Shield H,
Virus prefilter, Viresolve
®
Pro Shield H, Magnus 2.1 format, filtration area 0.51 m²
Expand
View Pricing