
Uracil arabinoside

Synonym(s):
Uracil 1-β-D-arabinofuranoside, 1-β-D-Arabinofuranosyluracil
Empirical Formula (Hill Notation):
C9H12N2O6
CAS Number:
Molecular Weight:
244.20
Products
Brand
Formula Weight
Manufacturer
Agency
Format
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade