Skip to Content
AM
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Tweezers
Synonym(s):
Tweezer
Products
Product Category
tweezers (13)
Brand
Sigma-Aldrich (4)
Manufacturer
Package Size
Filter Products
Showing 1-17 of 17
Compare
Product Number
Description
Pricing
T5540
Tweezers,
Style #3, very sharp, PTFE-coated, anti-magnetic stainless steel
Expand
View Pricing
T4537
Tweezers,
Style #5, Fine needle-sharp, anti-magnetic stainless steel
Expand
View Pricing
T4662
Tweezers,
Style #5, Fine needle-sharp, Inox alloy (not anti-magnetic)
Expand
View Pricing
T4287
Tweezers,
Style #3C, very sharp, anti-magnetic stainless steel
Expand
View Pricing
T5415
Tweezers,
Style #00, sharp, anti-magnetic stainless steel
Expand
View Pricing
T5790
Tweezers,
Style #4, needle-sharp, anti-magnetic stainless steel
Expand
View Pricing
932787
Tweezers,
Style 5TTH
Expand
View Pricing
T6040
Tweezers,
Style #5, Fine needle-sharp, PTFE-coated, anti-magnetic stainless steel
Expand
View Pricing
T5665
Tweezers,
Style #3C, very sharp, PTFE-coated, anti-magnetic stainless steel
Expand
View Pricing
T6165
Tweezers,
Style #7, sharp, hooked, PTFE-coated, anti-magnetic stainless steel
Expand
View Pricing
933066
Tweezers,
Style 3F
Expand
View Pricing
T4912
Tweezers,
Style #7, sharp, hooked, anti-magnetic stainless steel
Expand
View Pricing
932795
Tweezers,
Style 2AB, ESD Rubber Coated
Expand
View Pricing
T4412
Tweezers,
Style #5, Fine needle-sharp, non-magnetic alloy (tips may be restraightened when bent)
Expand
View Pricing
932868
Tweezers,
Style AA, ESD Rubber Coated Boley
Expand
View Pricing
T4162
Tweezers,
Style #3C, very sharp, non-magnetic alloy (tips may be restraightened when bent)
Expand
View Pricing
T4787
Tweezers,
Style #7, sharp, hooked, non-magnetic alloy (tips may be restraightened when bent)
Expand
View Pricing