
TSKgel® Butyl-NPR HPLC Column

Synonym(s):
TSKgel® Butyl-NPR HPLC Column
Products
Product Category
Manufacturer
Product Line
Separation Technique
ID (mm)
Length (cm)
Particle Size (µm)
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing