Skip to Content
AM
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
tri-Sodium citrate dihydrate
Synonym(s):
Sodium citrate tribasic dihydrate, Citric acid trisodium salt dihydrate, Trisodium citrate dihydrate
Linear Formula:
HOC(COONa)(CH
2
COONa)
2
· 2H
2
O
CAS Number:
6132-04-3
Molecular Weight:
294.10
EC Number:
200-675-3
Products
Product Category
salts (3)
buffers (1)
Brand
SAFC (3)
Sigma-Aldrich (1)
Feature
Formula Weight
Melting Point (°C)
Physical Form
Product Line
Agency
Quality Segment
Application
Markush Class
Markush Group
Filter Products
Showing 1-4 of 4
Compare
Product Number
Description
Pricing
1.06432
tri-Sodium citrate dihydrate,
crystalline, EMPROVE
®
ESSENTIAL, Ph. Eur., BP, JP, USP, E 331
Expand
View Pricing
1.06447
tri-Sodium citrate dihydrate,
EMPROVE
®
EXPERT, Ph. Eur., BP, ChP, JP, USP, ACS, E 331
Expand
View Pricing
1.06446
tri-Sodium citrate dihydrate,
powder, EMPROVE
®
ESSENTIAL, Ph. Eur., BP, JP, USP, E 331
Expand
View Pricing
S1804
tri-Sodium citrate dihydrate,
meets USP testing specifications
Expand
View Pricing