
Tris(hydroxymethyl)aminomethane

Synonym(s):
2-Amino-2-(hydroxymethyl)-1,3-propanediol, THAM, Tris base, Trometamol
Linear Formula:
NH2C(CH2OH)3
CAS Number:
Molecular Weight:
121.14
Products
Product Category
Brand
Boiling Point (°C)
Formula Weight
Melting Point (°C)
pH Value
Physical Form
pK Value
Product Line
Agency
Quality Segment
Application
Showing 1-2 of 2