Skip to Content
Merck
Tris(1-pyrrolidinyl)phosphine 97%

Tris(1-pyrrolidinyl)phosphine

Tris(1-pyrrolidinyl)phosphine 97%
Synonym(s):
Tripyrrolidinophosphine, Phosphorous acid tripyrrolidide, Tris(N,N-tetramethylene)phosphorous acid triamide
Empirical Formula (Hill Notation):
C12H24N3P
CAS Number:
Molecular Weight:
241.31

Products

Boiling Point (°C)
Formula Weight
Physical Form
Purity
Functional Group
Markush Class
Markush Group
Reaction Type
Reagent Type
Showing 1-1 of 1
Compare
Product Number
Description
Pricing