Skip to Content
Merck
Trichloroacetic acid ACS reagent, for the determination of Fe in blood according to Heilmeyer, ≥99.5%

Trichloroacetic acid

Trichloroacetic acid ACS reagent, for the determination of Fe in blood according to Heilmeyer, ≥99.5%
Synonym(s):
Trichloroacetic acid, TCA
Linear Formula:
Cl3CCOOH
CAS Number:
Molecular Weight:
163.39
EC Number:

Products

Product Category
Brand
Boiling Point (°C)
Color
Feature
Formula Weight
Melting Point (°C)
pH Value
Physical Form
Product Line
Purity
Agency
Quality Segment
Functional Group
Application
Markush Class
Markush Group
Special Grade
Technique