Skip to Content
AM
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Toluene-3,4-dithiol
Synonym(s):
3,4-Dimercaptotoluene, 4-Methyl-1,2-benzenedithiol, ‘Dithiol’
Linear Formula:
CH
3
C
6
H
3
(SH)
2
CAS Number:
496-74-2
Molecular Weight:
156.27
EC Number:
207-828-3
Products
Product Category
building blocks (1)
Brand
Sigma-Aldrich (1)
Supelco (1)
Boiling Point (°C)
Formula Weight
Melting Point (°C)
Physical Form
Purity
Quality Segment
Markush Class
Markush Group
Special Grade
Technical (1)
Technique
Filter Products
Showing 1-2 of 2
Compare
Product Number
Description
Pricing
D129208
Toluene-3,4-dithiol,
technical grade, 90%
Expand
View Pricing
89700
Toluene-3,4-dithiol,
for spectrophotometric det. of Mo, Sn, W, and also Ag and Re, ≥97.0%
Expand
View Pricing