Skip to Content
Merck
Thallium(III) nitrate trihydrate

Thallium(III) nitrate trihydrate

Thallium(III) nitrate trihydrate
Synonym(s):
TTN, Thallium trinitrate
Linear Formula:
Tl(NO3)3 · 3H2O
CAS Number:
Molecular Weight:
444.44
EC Number:

Products




Showing 1-1 of 1
Compare
Product Number
Description
Pricing