Skip to Content
Merck
Tetrakis(triphenylphosphine)platinum(0) 97%

Tetrakis(triphenylphosphine)platinum(0)

Tetrakis(triphenylphosphine)platinum(0) 97%
Synonym(s):
Platinum(0)-tetrakis(triphenylphosphine)
Linear Formula:
Pt[(C6H5)3P]4
CAS Number:
Molecular Weight:
1244.22
EC Number:

Products

Product Category
Brand
Formula Weight
Purity
Markush Class
Markush Group
Reagent Type
Core
Showing 1-1 of 1
Compare
Product Number
Description
Pricing