Skip to Content
AM
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Supelclean
™
Florisil
®
/Na
2
SO
4
SPE Tube
Products
Product Category
tubes (2)
Brand
Supelco (2)
Product Line
Agency
Chemical Composition
Separation Technique
Volume (ml)
Application
Package Size
Technique
Filter Products
Showing 1-2 of 2
Compare
Product Number
Description
Pricing
54116-U
Supelclean
™
Florisil
®
/Na
2
SO
4
SPE Tube,
polypropylene hardware, PE frit, bed A 2 g (Florisil
®
), bed B 2 g (Na
2
SO
4
), volume 6 mL, pk of 48
Expand
View Pricing
52582-U
Supelclean
™
Florisil
®
/Na
2
SO
4
SPE Tube,
glass hardware, PTFE frit, bed A 2 g (Florisil
®
), bed B 2 g (Na
2
SO
4
), volume 6 mL, pk of 48
Expand
View Pricing