
Sulfamethazine-(phenyl-13C6) hemihydrate

Synonym(s):
4-Amino-N-(4,6-dimethyl-2-pyrimidinyl)benzene-13C6-sulfonamide, Sulfadimidine-phenyl-13C6
Empirical Formula (Hill Notation):
13C6C6H14N4O2S · 0.5H2O
CAS Number:
Molecular Weight:
293.29
Products
Product Category
Formula Weight
Product Line
Format
Quality Segment
Application
Reference Material Type
Special Grade
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing