Skip to Content
AM
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Stainless steel - AISI 301
Products
Manufacturer
Physical Form
Filter Products
Showing 1-10 of 10
Compare
Product Number
Description
Pricing
GF63606090
Stainless steel - AISI 301,
foil, thickness 1 mm, Fe/Cr17%/Ni 7%/C 0.1max%, size 150 × 150 mm, temper hard
Expand
View Pricing
GF77628370
Stainless steel - AISI 301,
foil, thickness 1 mm, Fe/Cr17%/Ni 7%/C 0.1max%, size 25 × 25 mm, temper hard
Expand
View Pricing
GF90624180
Stainless steel - AISI 301,
foil, thickness 0.5 mm, Fe/Cr17%/Ni 7%/C 0.1max%, size 25 × 25 mm, temper hard
Expand
View Pricing
GF44600510
Stainless steel - AISI 301,
foil, Fe/Cr 17%/Ni 7%/C 0.1%max, thickness 1 mm, size 50 × 50 mm, temper hard
Expand
View Pricing
GF09903764
Stainless steel - AISI 301,
foil, Fe/Cr 17%/Ni 7%/C 0.1%max, thickness 0.5 mm, size 50 × 50 mm, temper hard
Expand
View Pricing
GF64598596
Stainless steel - AISI 301,
foil, Fe/Cr17%/Ni 7%/C 0.1max%, thickness 1 mm, size 300 × 300 mm, temper hard
Expand
View Pricing
GF02230535
Stainless steel - AISI 301,
foil, Fe/Cr 17%/Ni 7%/C 0.1%max, thickness 0.5 mm, size 300 × 300 mm, temper hard
Expand
View Pricing
GF79526649
Stainless steel - AISI 301,
foil, thickness 1 mm, Fe/Cr17%/Ni 7%/C 0.1max%, size 100 × 100 mm, temper hard
Expand
View Pricing
GF20422055
Stainless steel - AISI 301,
foil, Fe/Cr 17%/Ni 7%/C 0.1%max, thickness 0.5 mm, size 100 × 100 mm, temper hard
Expand
View Pricing
GF44549400
Stainless steel - AISI 301,
foil, Fe/Cr 17%/Ni 7%/C 0.1%max, thickness 0.5 mm, size 150 × 150 mm, temper hard
Expand
View Pricing