Skip to Content
Merck
(S,S)-DIPAMP 95%

(S,S)-DIPAMP

(S,S)-DIPAMP 95%
Synonym(s):
(1S,2S)-(+)-Bis[(2-methoxyphenyl)phenylphosphino]ethane, (S,S)-1,2-Bis[(2-methoxyphenyl)(phenylphosphino)]ethane, (S,S)-1,2-Ethanediylbis[(2-methoxyphenyl)phenylphosphine]
Linear Formula:
[CH3OC6H4P(C6H5)CH2]2
CAS Number:
Molecular Weight:
458.47

Products

Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Quality Segment
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing