
Sophocarpine

Empirical Formula (Hill Notation):
C15H22N2O
CAS Number:
Molecular Weight:
246.35
Products
Product Category
Brand
Biological Source
Color
Formula Weight
Manufacturer
Physical Form
Product Line
Purity
Format
Research Area
Reference Material Type
Special Grade
Showing 1-2 of 2
Compare
Product Number
Description
Pricing