
Sodium tungstate dihydrate

Synonym(s):
Tungstic acid sodium salt dihydrate, Sodium tungstate dihydrate
Linear Formula:
Na2WO4 · 2H2O
CAS Number:
Molecular Weight:
329.85
Products
Product Category
Brand
Formula Weight
Melting Point (°C)
pH Value
Physical Form
Product Line
Purity
Quality Segment
Reagent Type
Core
Special Grade
Showing 1-5 of 5
Compare
Product Number
Description
Pricing