Skip to Content
AM
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Sodium (meta)periodate
Synonym(s):
Sodium periodate, Sodium metaperiodate
Linear Formula:
NaIO
4
CAS Number:
7790-28-5
Molecular Weight:
213.89
EC Number:
232-197-6
Products
Product Category
salts (2)
Brand
Sigma-Aldrich (2)
Supelco (1)
Color
Formula Weight
pH Value
Physical Form
Product Line
Purity
Agency
Quality Segment
Reagent Type
Filter Products
Showing 1-4 of 4
Compare
Product Number
Description
Pricing
S1878
Sodium (meta)periodate,
≥99.0%
Expand
View Pricing
1.06597
Sodium (meta)periodate,
for analysis EMSURE
®
ACS,Reag. Ph Eur
Expand
View Pricing
769517
Sodium (meta)periodate,
≥99%
Expand
View Pricing
71859
Sodium (meta)periodate,
BioUltra, ≥99.5% (RT)
Expand
View Pricing