Skip to Content
Merck
Sodium cholate hydrate BioReagent, suitable for cell culture

Sodium cholate hydrate

Sodium cholate hydrate BioReagent, suitable for cell culture
Synonym(s):
3α,7α,12α-Trihydroxy-5β-cholan-24-oic acid sodium salt, Cholalic acid sodium salt
Empirical Formula (Hill Notation):
C24H39NaO5 · xH2O
CAS Number:
Molecular Weight:
430.55 (anhydrous basis)
EC Number:

Products

Product Category
Brand
Biological Source
Formula Weight
Physical Form
Product Line
Purity
Tissue
Quality Segment
Special Grade
Technique
Showing 1-5 of 5
Compare
Product Number
Description
Pricing