Skip to Content
AM
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Water Purification
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Sodium acetate buffer solution
CAS Number:
126-96-5
Products
Product Category
buffers (2)
Brand
Sigma-Aldrich (1)
pH Value
Physical Form
Product Line
Quality Segment
Sterility
Sterilization Method
Filter Products
Showing 1-2 of 2
Compare
Product Number
Description
Pricing
S7899
Sodium acetate buffer solution,
pH 5.2±0.1 (25 °C), Molecular Biology, 3 M, 0.2 μm filtered
Expand
View Pricing
S2404
Sodium acetate buffer solution,
BioXtra, pH 7.0±0.05 (25 °C), Molecular Biology, 3 M, non-sterile; 0.2 μm filtered
Expand
View Pricing