Skip to Content
AM
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Silica
Products
Brand
Sigma-Aldrich (5)
Physical Form
Diameter (mm)
Quality Segment
Functional Group
Particle Size (µm)
Filter Products
Showing 1-5 of 5
Compare
Product Number
Description
Pricing
900769
Silica,
mesoporous MCM-48, <15 μm particle size, pore size 3 nm, amine functionalized
Expand
View Pricing
900783
Silica,
mesoporous MCM-48, <15 μm particle size, pore size 3 nm, thiol functionalized
Expand
View Pricing
900780
Silica,
mesoporous SBA-15, <150 μm particle size, pore size 6 nm, amine functionalized
Expand
View Pricing
900773
Silica,
MCM-41, pellet, diam. × thickness 2 mm × 5 mm
Expand
View Pricing
900775
Silica,
SBA-15, pellet, diam. × thickness 2 mm × 5 mm
Expand
View Pricing