Skip to Content
AM
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Water Purification
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Sigma Aldrich
®
Dual Run & Blot Accessory
Products
Brand
Sigma-Aldrich (3)
Manufacturer
Length (cm)
Color
Plate Material
Package Size
Filter Products
Showing 1-4 of 4
Compare
Product Number
Description
Pricing
Z741772
Sigma Aldrich
®
Dual Run & Blot Accessory,
Sponge Pad Kit, size 9 cm × 9 cm, pack of 4 ea
Expand
View Pricing
Z741770
Sigma Aldrich
®
Dual Run & Blot Accessory,
Cooling Block
Expand
View Pricing
Z741769
Sigma Aldrich
®
Dual Run & Blot Accessory,
Blotting Cassette with Pad, L × W 9 cm × 9 cm
Expand
View Pricing
Z741771
Sigma Aldrich
®
Dual Run & Blot Accessory,
Buffer Dam
Expand
View Pricing