(S)-(+)-Mandelic acid
Synonym(s):
(S)-α-Hydroxyphenylacetic acid
Linear Formula:
C6H5CH(OH)CO2H
Products
Product Category
Brand
Formula Weight
Melting Point (°C)
Physical Form
Product Line
Purity
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing