
(S)-(+)-α-Methoxy-α-(trifluoromethyl)phenylacetyl chloride

Synonym(s):
(S)-(+)-MTPA-Cl, Mosher’s acid chloride
Linear Formula:
C6H5C(OCH3)(CF3)COCl
CAS Number:
Molecular Weight:
252.62
Products
Product Category
Brand
Boiling Point (°C)
Formula Weight
Physical Form
Purity
Quality Segment
Functional Group
Markush Class
Markush Group
Technique
Showing 1-2 of 2