
Risedronate sodium

Synonym(s):
Risedronic acid monosodium salt hemi(pentahydrate)
Empirical Formula (Hill Notation):
C7H10NNaO7P2 · 2.5H2O
CAS Number:
Molecular Weight:
350.13
Products
Brand
Formula Weight
Manufacturer
Physical Form
Agency
Format
Quality Segment
Application
Package Size
API Family
Special Grade
Showing 1-2 of 2