
Residual Solvent Class 2 - Methanol

Synonym(s):
Methanol solution
Empirical Formula (Hill Notation):
CH4O
CAS Number:
Molecular Weight:
32.04
Products
Formula Weight
Manufacturer
Format
Application
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing