
Racemethionine

Synonym(s):
DL-Methionine, (±)-2-Amino-4-(methylmercapto)butyric acid, DL-2-Amino-4-(methylthio)butanoic acid
Linear Formula:
CH3SCH2CH2CH(NH2)COOH
CAS Number:
Molecular Weight:
149.21
Products
Brand
Formula Weight
Manufacturer
Melting Point (°C)
Physical Form
Agency
Format
Quality Segment
Application
API Family
Reference Material Type
Special Grade
Showing 1-2 of 2