Skip to Content
AM
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Water Purification
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Quickfit
®
flat flange lids for reaction flasks
Products
Product Category
lids (8)
Manufacturer
Package Size
Sterility
Filter Products
Showing 1-8 of 8
Compare
Product Number
Description
Pricing
Z303887
Quickfit
®
flat flange lids for reaction flasks,
center joint: ST/NS 29/32
Expand
View Pricing
Z303933
Quickfit
®
flat flange lids for reaction flasks,
center joint: ST/NS 24/29, 5° side socket joint: 19/26, parallel socket joint: 19/26, 10° side socket joint: 19/26, 15° side socket joint: 34/35
Expand
View Pricing
Z691046
Quickfit
®
flat flange lids for reaction flasks,
flange bore 75 mm, center joint: ST/NS 19/26
Expand
View Pricing
Z691038
Quickfit
®
flat flange lids for reaction flasks,
flange bore 75 mm, center joint: ST/NS 34/35
Expand
View Pricing
Z309060
Quickfit
®
flat flange lids for reaction flasks,
center joint: ST/NS 19/26, parallel socket joint: 19/26, 5° side socket joint: 19/26, 10° side socket joint: 19/26, 15° side socket joint: 34/35
Expand
View Pricing
Z309052
Quickfit
®
flat flange lids for reaction flasks,
center joint: ST/NS 19/26, parallel socket joint: 19/26, 5° side socket Joints: 19/26 (2), 10° side socket joint: 19/26
Expand
View Pricing
Z309079
Quickfit
®
flat flange lids for reaction flasks,
center joint: ST/NS 29/32, parallel socket joint: 14/23, 5° side socket joint: 14/23, 10° side socket joint: 14/23, 15° side socket joint: 29/32
Expand
View Pricing
Z303909
Quickfit
®
flat flange lids for reaction flasks,
center joint: ST/NS 19/26, parallel socket joint: 19/26, 5° side socket joint: 24/29, 10° side socket joint: 19/26
Expand
View Pricing