Skip to Content
AM
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Pyrex
®
Measuring cylinder, class B, squat form, heavy duty rim, blue graduations
Products
Product Category
cylinders (5)
Manufacturer
Color
Filter Products
Showing 1-5 of 5
Compare
Product Number
Description
Pricing
SLW3285-100
Pyrex
®
Measuring cylinder, class B, squat form, heavy duty rim, blue graduations,
10 mL
Expand
View Pricing
SLW3285-500
Pyrex
®
Measuring cylinder, class B, squat form, heavy duty rim, blue graduations,
500 mL
Expand
View Pricing
SLW3285-25
Pyrex
®
Measuring cylinder, class B, squat form, heavy duty rim, blue graduations,
25 mL
Expand
View Pricing
SLW3285-10
Pyrex
®
Measuring cylinder, class B, squat form, heavy duty rim, blue graduations,
10 mL
Expand
View Pricing
SLW3285-50
Pyrex
®
Measuring cylinder, class B, squat form, heavy duty rim, blue graduations,
50 mL
Expand
View Pricing